Skip to Content
Merck
Benzyl cinnamate phyproof® Reference Substance

Benzyl cinnamate

Benzyl cinnamate phyproof® Reference Substance
Synonym(s):
3-Phenyl 2-propenoic acid benzyl ester, Benzyl 3-phenylpropenoate, Cinnamic acid benzyl ester
Linear Formula:
C6H5CH=CHCOOCH2C6H5
CAS Number:
Molecular Weight:
238.28

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade