
Atrazine-desisopropyl

Synonym(s):
2-Amino-4-chloro-6-ethylamino-1,3,5-triazine
Empirical Formula (Hill Notation):
C5H8ClN5
CAS Number:
Molecular Weight:
173.60
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing