Skip to Content
Merck
Ampliflu™ Red for fluorescence, ≥98.0% (HPLC)

Ampliflu Red

Ampliflu™ Red for fluorescence, ≥98.0% (HPLC)
Synonym(s):
10-Acetyl-3,7-dihydroxyphenoxazine
Empirical Formula (Hill Notation):
C14H11NO4
CAS Number:
Molecular Weight:
257.24

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing