
Ammonium trifluoroacetate

Synonym(s):
Trifluoroacetic acid ammonium salt
Linear Formula:
CF3CO2NH4
CAS Number:
Molecular Weight:
131.05
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing