
Ammonium-d4 deuteroxide solution

Empirical Formula (Hill Notation):
ND4OD
CAS Number:
Molecular Weight:
40.08
Products
Product Category
Formula Weight
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing