Skip to Content
Merck
Acrylic acid N-hydroxysuccinimide ester ≥90% (GC)

Acrylic acid N-hydroxysuccinimide ester

Acrylic acid N-hydroxysuccinimide ester ≥90% (GC)
Synonym(s):
N-Acryloxysuccinimide
Empirical Formula (Hill Notation):
C7H7NO4
CAS Number:
Molecular Weight:
169.13

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing