
Acetylsalicylic acid

Synonym(s):
2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin
Linear Formula:
2-(CH3CO2)C6H4CO2H
CAS Number:
Molecular Weight:
180.16
Products
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
pK Value
Purity
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing