
4-Hydroxycoumarin

Synonym(s):
4-Hydroxy-1-benzopyran-2-one
Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing