Skip to Content
Merck
4-(Diethylamino)salicylaldehyde 98%

4-(Diethylamino)salicylaldehyde

4-(Diethylamino)salicylaldehyde 98%
Synonym(s):
4-Diethylamino-2-hydroxybenzaldehyde
Linear Formula:
(C2H5)2NC6H3-2-(OH)CHO
CAS Number:
Molecular Weight:
193.24
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing