Skip to Content
Merck
3-Thiopheneethanol 99%

3-Thiopheneethanol

3-Thiopheneethanol 99%
Synonym(s):
2-(3-Thienyl)ethanol, 3-(2-Hydroxyethyl)thiophene
Empirical Formula (Hill Notation):
C6H8OS
CAS Number:
Molecular Weight:
128.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing