
3,3′-Dithiodipropionic acid di(N-hydroxysuccinimide ester)

Synonym(s):
DTSP, Di(N-succinimidyl) 3,3′-dithiodipropionate, Dithiobis(succinimidyl propionate), Lomant’s reagent
Empirical Formula (Hill Notation):
C14H16N2O8S2
Products
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing