
3,3′-Diethylthiacarbocyanine iodide

Synonym(s):
3-Ethyl-2-(3-[3-ethyl-2(3H)benzothiazolylidene]-1-propenyl)benzothiazolium iodide, DiSC2(3)
Empirical Formula (Hill Notation):
C21H21IN2S2
CAS Number:
Molecular Weight:
492.44
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing