Skip to Content
Merck
vibrant-m

2,5-Dimethyl-2,5-dihydroxy-1,4-dithiane

vibrant-m
Synonym(s):
1-Mercaptopropanone (dimer), 2,5-Dihydroxy-2,5-dimethyl-1,4-dithiane, 2,5-Dimethyl-1,4-dithiane-2,5-diol, 2,5-Dimethyl-2,5-dihydroxy-p-dithiane, Cyclodithalfarol-705
Empirical Formula (Hill Notation):
C6H12O2S2
CAS Number:
Molecular Weight:
180.29
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing