
2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate

Synonym(s):
4-Methylumbelliferyl-N-acetyl-α-D-neuraminic acid sodium salt hydrate
Empirical Formula (Hill Notation):
C21H24NNaO11 · xH2O
CAS Number:
Molecular Weight:
489.41 (anhydrous basis)
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment