Skip to Content
Merck
1-Butanol anhydrous, ZerO2®, 99.8%

1-Butanol

1-Butanol anhydrous, ZerO2®, 99.8%
Synonym(s):
n-Butanol, Butyl alcohol, 1-Butanol
Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique