Skip to Content
Merck
1-Bromo-3,5-di-tert-butylbenzene 97%

1-Bromo-3,5-di-tert-butylbenzene

1-Bromo-3,5-di-tert-butylbenzene 97%
Linear Formula:
BrC6H3(C(CH3)3)2
CAS Number:
Molecular Weight:
269.22

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing