Amino Acids
Amino acids (AAs) are a group of organic molecules in which each is comprised of a basic amino group (-NH2), an acidic carboxyl group (-COOH), and an organic R group (or side chain) that is unique to each amino acid. Amino acids are building blocks of proteins, as well as the intermediates in metabolism. The amino and the carboxyl groups of amino acids react to form a covalent amide linkage, called a peptide bond. This feature of amino acids allows them to polymerize to form proteins as well as peptides, which generally have shorter amino acid chain length. Amino acids can be classified into three groups: essential amino acids, non-essential amino acids and conditional amino acids. Amino acids are commonly used as supplements in cell culture media and in metabolism research. Discover how our amino acids in varying purities and grades can make a difference in your research endeavors.
Section Overview
Related Product Resources
- Amino Acids Reference Chart
Amino acid reference chart and products cater to diverse eukaryotic needs.
- Unnatural Amino Acids for Peptide Synthesis
Unnatural amino acids, the non-proteinogenic amino acids that either occur naturally or are chemically synthesized, are becoming more and more important as tools for modern drug discovery research.
- Amino Acid Codon Wheel
Amino Acid Codon Wheel for fast RNA translation. Find which amino acid is translated from your RNA sequence quickly and easily.
- L-Glutamine in Cell Culture
Importance and uses of glutamine in hybridoma and mammalian cell culture
Amino Acids in Cell Culture
As the building blocks of proteins, amino acids are critical for inclusion in cell culture media. Non-essential amino acids (NEAA) are those that are typically made by the organism, but can be added to culture media — individually or in the form of an NEAA cocktail — as a supplement to stimulate cell growth and promote longevity.
Polyamino Acids
Our polyamino acids have properties that mimic proteins, making them ideal for both drug delivery and the delivery of nucleic acids both in vitro and in vivo. We supply both research and cGMP polyamino acids with improved solubility, drug attachment stability, drug encapsulation, drug targeting, bypassing multidrug resistance (MDR) factors, minimal stimulation of the immune system, low toxicity, and biodegradability. We manufacture custom polyamino acids for specific molecular weight ranges or polydispersity ranges (size distribution). Moreover, we can conjugate polyamino acids to active pharmaceutical ingredients (API) for your drug delivery use.
Unnatural Amino Acids
Unnatural amino acids are non-proteinogenic amino acids that either occur naturally or are chemically synthesized. Whether utilized as building blocks, conformational constraints, molecular scaffolds, or pharmacologically active products, our unnatural amino acids represent a nearly infinite array of diverse structural elements for the development of new leads in peptidic and non-peptidic compounds. Due to their seemingly unlimited structural diversity and functional versatility, they are widely used as chiral building blocks and molecular scaffolds in constructing combinatorial libraries. Used as molecular probes, they can help to better understand the function of biological systems. Optimized and fine-tuned analogues of peptidic substrates, inhibitors, or effectors are also excellent analytical tools and molecular probes for investigating signal transduction pathways or gene regulation.