Skip to Content
Merck
All Photos(1)

Key Documents

PHL22470

Dillapiole

phyproof® Reference Substance

Synonym(s):

6-Allyl-4,5-dimethoxy-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24
Beilstein:
196037
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (GC)

manufacturer/tradename

PhytoLab

technique(s)

HPLC: suitable

density

1.163 g/mL at 20 °C (lit.)

application(s)

food and beverages
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

COc1c(CC=C)cc2OCOc2c1OC

InChI

1S/C12H14O4/c1-4-5-8-6-9-11(16-7-15-9)12(14-3)10(8)13-2/h4,6H,1,5,7H2,2-3H3

InChI key

LIKYNOPXHGPMIH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Packaging

This reference substance will be packed in micro-volume insert vials.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service