Skip to Content
Merck
All Photos(1)

Key Documents

814947

TSKgel® Butyl-NPR HPLC Column

phase C4 (butyl), L × I.D. 3.5 cm × 4.6 mm, 2.5 μm particle size

Synonym(s):

TSKgel® Butyl-NPR HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

material

PEEK

product line

TSKgel®

manufacturer/tradename

Tosoh Butyl-NPR, 14947

parameter

≤4 M salt concentration
≤50% organic solvent
1.2 mL/min flow rate
10-60 °C temperature
200 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

3.5 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

C4 (butyl) phase

particle size

2.5 μm

operating pH

2-12

separation technique

hydrophobic interaction (HIC)

Looking for similar products? Visit Product Comparison Guide

General description

The least hydrophobic of the HIC column offerings, TSKgel Butyl-NPR is the best choice for high speed separations with excellent recovery even for more hydrophobicity samples. The non-porous resin requires lower sample loading and leads to faster analysis time because the binding kinetics occur only on the bead′s surface. The ultra efficient 2.5 micron bead allows for fast, high efficiency separations using a 3.5cm x 4.6mm ID column format. Thus, TSKgel Butyl-NPR is ideal for time critical QC analysis or sample limited applications. Featuring polymethacrylate base material, the TSKgel Butyl-NPR column is stable in either acid or caustic cleaning regimes up to 1N.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service