Skip to Content
Merck
All Photos(1)

Key Documents

SML2466

Sigma-Aldrich

OXFBD04

≥98% (HPLC)

Synonym(s):

3-(3,5-dimethylisoxazol-4-yl)-5-(hydroxy(pyridin-3-yl)methyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2O3
CAS Number:
Molecular Weight:
296.32
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=NOC(C)=C1C2=CC(C(O)C3=CN=CC=C3)=CC(O)=C2

Biochem/physiol Actions

OXFBD04 is a potent and selective inhibitor of BRD4(1) that inhibits interactions of BRD4with the RelA subunit of NF-κB. OXFBD02 inhibits proliferation of various cancer cell lines including leukaemia, breast, and renal cancer cell lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura E Jennings et al.
Bioorganic & medicinal chemistry, 26(11), 2937-2957 (2018-05-20)
Ligands for the bromodomain and extra-terminal domain (BET) family of bromodomains have shown promise as useful therapeutic agents for treating a range of cancers and inflammation. Here we report that our previously developed 3,5-dimethylisoxazole-based BET bromodomain ligand (OXFBD02) inhibits interactions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service