Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702248

Sigma-Aldrich

Anti-Folate Binding Protein(FBP) / FOLR1 Antibody, clone 3J6W3, Rabbit Monoclonal

Synonym(s):

FOLR1,FBP,FOLR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

material

colorless

clone

3J6W3, monoclonal

form

liquid

mol wt

38 kDa

species reactivity

human, rat, rat

concentration

0.93 mg/mL

technique(s)

western blot: 1:500 - 1:1000

color

colorless

isotype

IgG

immunogen sequence

MAQRMTTQLLLLLVWVAVVGEAQTRIAWARTELLNVCMNAKHHKEKPGPEDKLHEQCRPWRKNACCSTNTSQEAHKDVSYLYRFNWNHCGEMAPACKRHF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOLR1(2348)

General description

The protein encoded by this gene is a member of the folate receptor family. Members of this gene family bind folic acid and its reduced derivatives, and transport 5-methyltetrahydrofolate into cells. This gene product is a secreted protein that either anchors to membranes via a glycosyl-phosphatidylinositol linkage or exists in a soluble form. Mutations in this gene have been associated with neurodegeneration due to cerebral folate transport deficiency. Due to the presence of two promoters, multiple transcription start sites, and alternative splicing, multiple transcript variants encoding the same protein have been found for this gene.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Folate Binding Protein(FBP) / FOLR1 (NP_000793.1).
PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service