Skip to Content
Merck
All Photos(1)

Key Documents

S3146

Sigma-Aldrich

Micro-dissecting scissors

size 4 1/4 in., straight, sharp point stainless steel

Synonym(s):

Scissors for Dissection

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122408
NACRES:
NB.35

material

straight, sharp point stainless steel

size

4 1/4 in.

Looking for similar products? Visit Product Comparison Guide

Application

Micro-dissecting scissorshave been used:

  • to decapitate mice embryos and to cut the limbs and tail
  • todissect the abdominal cavity and the testes of male mice
  • to cut open theabdominal cavity and peritoneum to expose embryo sacs from female mice

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolating Immune Cells from Mouse Embryonic Skin
Ambika S, et al.
Methods in Molecular Biology (2018)
Qianxing Zou et al.
STAR protocols, 3(2), 101398-101398 (2022-05-24)
Spermatogenesis generates heterologous cell populations which, if not distinguished clearly, often hinder mechanistic and etiological studies. Here, we present a protocol to identify and isolate populations of mouse spermatogenic cells, including spermatogonial stem cells (SSCs), spermatocytes, and haploid spermatids. We
Fábio F Rosa et al.
Bio-protocol, 10(10), e3619-e3619 (2021-03-05)
Ectopic expression of transcription factor combinations has been recently demonstrated to reprogram differentiated somatic cells towards the dendritic cell (DC) lineage without reversion to a multipotent state. DCs have the ability to induce potent and long-lasting adaptive immune responses. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service