Skip to Content
Merck
All Photos(1)

Key Documents

S0577

Sigma-Aldrich

Streptokinase C from Streptococcus sp.

lyophilized solid, ≥3,500 unit/mg solid

Synonym(s):

Streptokinase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Streptococcus sp.)

Quality Level

recombinant

expressed in E. coli

Assay

40—60% protein (biuret)

form

lyophilized solid

specific activity

≥3,500 unit/mg solid

color

off-white to light yellow

application(s)

life science and biopharma

storage temp.

−20°C

Gene Information

Streptococcus pyogenes ... SKC(134997)

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Streptokinase is an extracellular enzyme, a single-chain polypeptide produced by several strains of β-hemolytic Streptococci. It is composed of multiple structural domains like α-, β-, and γ-domains that are involved in various functions.

Application

Streptokinase C from Streptococcus sp. has been used to quantitate the survival of Staphylococcus aureus in whole human blood.

Biochem/physiol Actions

Streptokinase shows fibrinolytic activity by triggering the circulatory plasminogen either via a fibrin-dependent or fibrin-independent mechanism. Streptokinase has been studied majorly in terms of its therapeutic efficacy and treating myocardial infarction to dissolve clots.

Unit Definition

One unit will liquify a standard clot of fibrinogen, plasminogen, and thrombin at pH 7.5 at 37 °C in 10 min.

Physical form

Lyophilized powder containing approx. 50% bovine serum albumin (total protein by biuret) with sodium glutamate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerstin Steiner et al.
Infection and immunity, 70(7), 3627-3636 (2002-06-18)
Synthesis of the plasminogen activator streptokinase (SK) by group A streptococci (GAS) has recently been shown to be subject to control by two two-component regulators, covRS (or csrRS) and fasBCA. In independent studies, response regulator CovR proved to act as
Streptokinase production from Streptococcus dysgalactiae subsp. equisimilis SK-6 in the presence of surfactants, growth factors and trace elements
Bhardwaj S and Angayarkann J
3 Biotech, 5(2), 187?193-187?193 (2015)
Jakub M Kwiecinski et al.
Cell reports, 36(4), 109462-109462 (2021-07-29)
Skin is one of the most common sites of host immune response against Staphylococcus aureus infection. Here, through a combination of in vitro assays, mouse models, and intravital imaging, we find that S. aureus immune evasion in skin is controlled by a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service