Skip to Content
Merck
All Photos(2)

Key Documents

RAB0035

Sigma-Aldrich

Human Carbohydrate Antigen 19-9 ELISA Kit

for serum, plasma and cell culture supernatants

Synonym(s):

CA19-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

technique(s)

ELISA: suitable
capture ELISA: suitable

shipped in

wet ice

storage temp.

−20°C

General description

The Human CA19-9 (Cancer Antigen 19-9) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CA19-9 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human CA19-9

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Cancer antigen 19-9 or carbohydrate antigen 19-9 is synthesized by pancreatic and biliary ductular cells, as well as by salivary, colonic, gastric, endometrial epithelia. It is also found in seminal fluid. In tissues, CA 19-9 exists as an epitope present on a glycolipid, whereas in serum, it is associated with mucin. CA 19-9 is a marker for pancreatic, gastric and hepatobiliary malignancies and is found in elevated levels.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The clinical significance of elevated levels of serum CA 19-9.
Pavai S and Yap SF
The Medical Journal of Malaysia, 58(5), 667-672 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service