Skip to Content
Merck
All Photos(1)

Key Documents

MTOXCE3A4

Sigma-Aldrich

CypExpress 3A4 Cytochrome P450 human

Synonym(s):

CP33, CP34, CYP3A, CYP3A3, CYP3A4, CYPIIIA3, CYPIIIA4, HLP, NF-25, P450 3A4, P450C3, P450PCN1

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106200
NACRES:
NA.77

biological source

human

recombinant

expressed in Pichia pastoris

form

dry powder

UniProt accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CYP3A4(1576)

Related Categories

Biochem/physiol Actions

Cytochrome P450 enzymes are heme containing monooxygenases which are found primarily in the mammalian liver. They catalyze the oxidative metabolism of xenobiotics. This metabolism is the initial step in the biotransformation and elimination of a wide variety of drugs and environmental pollutants from the body. Ketoconazole, itraconazole, azamulin, troleandomycin and verapamil are inhibitors, whereas rifampin is an inducer of the enzyme. The isozyme CYP3A4 is a major pathway for the invivo conversion of testosterone to 6β-Hydroxytestosterone via hydroxylation.
Cytochrome P450 is a heterogeneous family of isozymes whose primary function is to oxidize small molecules, both as a function of intermediary metabolism (e.g., fatty acids) and to detoxify exogenous compounds like xenobiotics, pollutants and drug molecules. Cytochrome P450 3A4 (CYP3A4) is one of the isoforms of cytochrome P450 (P450s). CYP3A4 oxidizes more than 90% of marketed drugs through various metabolic processes.

Features and Benefits

CypExpress 3A4 is a permeabilized and stabilized dried yeast powder preparation containing full length, unmodified, human CYP 3A4 and recombinant human NADPH oxidoreductase. Using proprietary processes, CypExpress are expressed in Pichia yeast. Following optimal expression, the cells are inactivated, permeabilized and dried to a fine powder. CypExpress retains the cellular mechanisms to provide the P450 enzymes with the energy and cofactors to continue to function for long experiments, and can generate larger amounts of metabolite than mammalian microsomes or other genetically engineered expression systems. It is designed for rapid screening of drug metabolites and for scaling up metabolite generation.

Unit Definition

A reaction mixture containing 500 μM testosterone and 100 mg/mL of CypExpress 3A4 will produce 121.9 μM 6b-hydroxytestosterone in 3 hours at 30°C under the conditions described.

Physical form

Light yellow, very fine powder

Reconstitution

Per Protocol/Variable

Legal Information

6538 is a trademark of American Type Culture Collection
CypExpress is a trademark of Oxford Biomedical Research

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structures of human cytochrome P450 3A4 bound to metyrapone and progesterone
Williams PA, et al.
Science, 305(5684), 683-686 (2004)
Pharmacokinetic-pharmacodynamic consequences and clinical relevance of cytochrome P450 3A4 inhibition
Dresser, et al.
Clinical Pharmacokinetics, 38(1), 41-57 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service