Skip to Content
Merck
All Photos(1)

Key Documents

M9773

Sigma-Aldrich

4-Methylbenzhydrylamine resin hydrochloride

50-200 mesh

Synonym(s):

(α-Amino-α-p-xylyl hydrochloride)polystyrene crosslinked with divinylbenzene, Amino-(4-methylphenyl)methyl copolystyrene divinylbenzene, MBHA-polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56

form

solid

extent of labeling

0.5-0.7 amine per g resin (based on nitrogen determination)

matrix

2% Cross-linked polystyrene

particle size

50-200 mesh

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Hydrochloride resins have been used to study the treatment of hyperphosphataemia in patients with end-stage renal failure.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Spaia
Hippokratia, 15(Suppl 1), 22-26 (2011-09-08)
In chronic kidney disease patients, bone and mineral abnormalities have a major impact on morbidity and mortality. Hyperphosphatemia has been associated with increased mortality and with the development of cardiovascular calcification, an independent predictor of mortality. Sevelamer, or more precisely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service