Skip to Content
Merck
All Photos(8)

Key Documents

HPA012778

Sigma-Aldrich

Anti-PPP3CA antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAM-PRP catalytic subunit, Anti-Calmodulin-dependent calcineurin A subunit alpha isoform, Anti-Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, rat, mouse

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SEPKAIDPKLSTTDRVVKAVPFPPSHRLTAKEVFDNDGKPRVDILKAHLMKEGRLEESVALRIITEGASILRQEKNLLDIDAPVTVCGDIHGQFFDLMKLFEVG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPP3CA(5530)

General description

Calmodulin-dependent calcineurin A subunit α isoform (PPP3CA) is a catalytic subunit of the cytosolic phosphatase calcineurin. The protein contains a calmodulin-binding site. The gene encoding the protein is present on chromosome 4.

Immunogen

Serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Calmodulin-dependent calcineurin A subunit α isoform (PPP3CA) is involved in the mediation of Ca2+-induced cofilin-phosphatase slingshot-1L (SSH1L) activation and cofilin dephosphorylation. It recognises substrates which contain a PXIXIT motif. The translocation of the dynamin related protein 1 (Drp1) is carried out by PPP3CA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71581

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiming Li et al.
Journal of molecular biology, 369(5), 1296-1306 (2007-05-15)
The protein phosphatase calcineurin recognizes a wide assortment of substrates and controls diverse developmental and physiological pathways in eukaryotic cells. Dephosphorylation of the transcription factor NFAT and certain other calcineurin substrates depends on docking of calcineurin at a PxIxIT consensus
Yan Wang et al.
The Journal of biological chemistry, 280(13), 12683-12689 (2005-01-27)
Cofilin, an essential regulator of actin filament dynamics, is inactivated by phosphorylation at Ser-3 and reactivated by dephosphorylation. Although cofilin undergoes dephosphorylation in response to extracellular stimuli that elevate intracellular Ca2+ concentrations, signaling mechanisms mediating Ca2+-induced cofilin dephosphorylation have remained
N Frey et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(26), 14632-14637 (2000-12-13)
The calcium- and calmodulin-dependent protein phosphatase calcineurin has been implicated in the transduction of signals that control the hypertrophy of cardiac muscle and slow fiber gene expression in skeletal muscle. To identify proteins that mediate the effects of calcineurin on
G M Cereghetti et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(41), 15803-15808 (2008-10-08)
Changes in mitochondrial morphology that occur during cell cycle, differentiation, and death are tightly regulated by the balance between fusion and fission processes. Excessive fragmentation can be caused by inhibition of the fusion machinery and is a common consequence of
M G Wang et al.
Cytogenetics and cell genetics, 72(2-3), 236-241 (1996-01-01)
Calcineurin (also called protein phosphatase-2B) is a calmodulin-regulated protein phosphatase which plays an important role in signal transduction. The enzyme is a heterodimer of a 58-59 kDa calmodulin-binding catalytic subunit (calcineurin A) and a small (i.e. 19 kDa) Ca(2+)-binding regulatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service