Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0031-01

Sephadex® G-25 Superfine

Cytiva 17-0031-01, pack of 100 g

Synonym(s):

Gel Filtration Media, Sephadex G-25

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

packaging

pack of 100 g

manufacturer/tradename

Cytiva 17-0031-01

storage condition

dry media

matrix

cross-linked dextran

particle size

>20 μm (dry)

cleaning in place

2-13

working range

2-13

suitability

suitable for bioprocess medium

General description

Sephadex® G-25 is well established gel filtration medium for desalting and buffer exchange in industrial applications.

Application

Sephadex® is a gel filtration medium prepared by crosslinking Dextran with epichlorohydrin. Different types of Sephadex® differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex® G-25 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex® G-25 is available in 4 different particle sizes (Course, Medium, Fine & Superfine).

Sephadex® G-25 is available in different bulk pack sizes and convenient pre-packed formats.
As a BioProcess medium, Sephadex® G-25, meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
  • Excellent recovery and minimum sample dilution
  • Available in prepacked HiPrep Desalting and HiTrap® Desalting columns for fast and convenient desalting
  • BioProcess medium supported for industrial applications and well-established for desalting and buffer exchange in industrial applications

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (4 to 8, Used Media 20% Ehtanol or 0.01 M NaOH)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiPrep is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
Sephadex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.

This page shows how to solve practical problems that may occur when running an affinity chromatography column.

Protocols

This page shows describes fractional precipitation for removing gross impurities from affinity chromatography samples with small volumes.

This page shows how to perform a separation with a Sephadex column from Cytiva which are Size Exclusion Chromatography media consisting of an epichlorohydrin cross-linked dextran.

How to perform buffer exchange and desalting with Sephadex G-25, HiTrap Desalting columns, or ÄKTAprime plus.

This page shows how to prepare samples for purification with affinity chromatography.

Related Content

Separation options for Size Exclusion Chromatography with Sephadex from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service