EP5948000810
Eppendorf® Centrifuge 5920 R G
AC/DC input 230 V, 50 - 60 Hz, incl. rotor S-4xuniversal large and adapter 13/16 mm round bottom tubes
About This Item
Recommended Products
AC/DC input
, Schuko plug
feature
CE compliant
refrigerated
to hold 148 16 mm round bottom tubes
to hold 196 13 mm round bottom tubes
to hold 4 universal buckets
incl. rotor S-4xuniversal large and adapter 13/16 mm round bottom tubes
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 5948000810 (EU)
parameter
temp. range at -11-40 °C
13700 rpm
21194 × g
technique(s)
centrifugation: suitable
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Swing-bucket rotors and adapters accommodate tubes and bottles from 0.2 mL to 1,000 mL
- Plate rotor options for centrifugation of all types of MTP, PCR or Deepwell Plates
- Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 50 mL
- High centrifugation speed of up to 21.194 × g (13,700 rpm)
- Aerosol-tight Eppendorf QuickLock caps and lids allow fast and ergonomic lid locking
- Centrifuge lid with soft-touch lid closure
- Quiet operation to improve your work environment
- Compact footprint saves valuable bench space
- Automatic rotor recognition and imbalance detection for maximum operational safety
- Temperature range from -11 °C to 40 °C
- FastTemp function for quick pre-cooling
- FastTemp pro function allows for automatic precooling based on pre-programmable time and date
- Continuous cooling maintains a constant temperature after the run has been completed – your samples stay cool
- ECO shut-off engages after 8 hours (adjustable) of non-use to reduce energy consumption and to extend compressor life
- Dynamic compressor control (DCC) technology for precise cooling performance
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service