Skip to Content
Merck
All Photos(1)

Documents

NP cl.25

Reactivity: Anti-Measles Virus Nucleoprotein, 95051114

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Mouse Hybridoma

ImmunoDon

BALB/c mouse spleen

Myeloma

Sp2/0 Ag-14

Reactivity

Anti-Measles Virus Nucleoprotein

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse Sp2/0 x Mouse BALB/c spleen

Cell Line Description

NP cl.25 monoclonal antibody was produced in vitro by fusing mouse myeloma cells with spleen cells derived from BALB/c mice immunised with purified measles virus. NP cl.25 is negative for haemagglutination and hemolysin inhibition as well as neutralisation assays and gives immunofluorescence only on fixed tissue. It has been used for measuring synthesis and topological accumulation of measles virus antigen during the lytic cycle.

Immunogen

Measles virus (strain Hallè)

Application

Diagnosis, study of measles virus infection

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 1-5x100,000 cells/ml; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service