Skip to Content
Merck
All Photos(1)

Key Documents

Do-11-10

85082301, mouse unknown/unspecified, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Do-11-10, from mouse unknown/unspecified, 85082301

biological source

mouse unknown/unspecified

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c (H-2d) T cells x Mouse AKR Thyoma, BW5147

Cell Line Description

Do-11-11 was produced by fusion of the antigen-specific I region-restricted (H-2d) BALB/c T-cell blasts with the AKR thyoma BW5147. This murine helper T cell hybridoma is specific for chicken ovalbumin (cOva) restricted to I-Ad and cross reacts with cOva I-antibody. The hybrid can be stimulated to secrete Interleukin 2 (IL-2) or to act as an antigen-specific helper T-cell. Expression of IL-2 can be lost by long-term culture of growth at high densities.

Culture Medium

DMEM or RPMI 1640 + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37 °C. Cells should not be grown at high density; avoid long-term culture as this might lead to change of cell receptors.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service