Skip to Content
Merck
All Photos(1)

Key Documents

73766

Sigma-Aldrich

Tetramethylrhodamine isothiocyanate–Dextran

average mol wt 20,000

Synonym(s):

TRITC-Dextran 20, TRITC-Dextran

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.32

biological source

synthetic

form

powder

mol wt

average mol wt 20,000

extent of labeling

0.001-0.008 mol/mol TRITC glucose

fluorescence

λex 550 nm; λem 577 nm±5 nm in 0.1 M Tris pH 8.0

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilie Le Maout et al.
Biophysical journal, 119(7), 1301-1308 (2020-10-08)
Cell motility is essential in a variety of biological phenomena ranging from early development to organ homeostasis and diseases. This phenomenon has mainly been studied and characterized on flat surfaces in vitro, whereas such conditions are rarely observed in vivo. Recently, cell

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service