Skip to Content
Merck
All Photos(1)

Key Documents

Y0000377

Ribavirin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-β-D-Ribofuranosyl-1,2,4-triazole-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N4O5
CAS Number:
Molecular Weight:
244.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ribavirin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)c1ncn(n1)[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O

InChI

1S/C8H12N4O5/c9-6(16)7-10-2-12(11-7)8-5(15)4(14)3(1-13)17-8/h2-5,8,13-15H,1H2,(H2,9,16)/t3-,4-,5-,8-/m1/s1

InChI key

IWUCXVSUMQZMFG-AFCXAGJDSA-N

Gene Information

human ... IMPDH1(3614)

Looking for similar products? Visit Product Comparison Guide

General description

Analog of the imidazole nucleotide intermediates in purine nucleotide biosynthesis.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ribavirin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Antiviral agent used against a wide variety of human viral infections, in particular, chronic hepatitis C, HIV, and adenovirus. Its metabolite, ribavirin 5′-phosphate, is an inhibitor of inosine monophosphate (IMP) dehydrogenase, but many other mechanisms of action are also supported with experimental evidence.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Muta. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tung-Hung Su et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(19), 7844-7849 (2013-04-25)
MicroRNA-122 (miR-122) facilitates hepatitis C virus replication in vitro. Serum miR-122 has been implicated as a biomarker for various liver diseases; however, its role in chronic hepatitis C remains unclear. To address this issue, 126 patients with chronic hepatitis C
Chen-Hua Liu et al.
Annals of internal medicine, 159(11), 729-738 (2013-12-04)
Data are limited on the efficacy and safety of pegylated interferon plus ribavirin for patients with hepatitis C virus genotype 1 (HCV-1) receiving hemodialysis. To compare the efficacy and safety of combination therapy with pegylated interferon plus low-dose ribavirin and
Eric Druyts et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 56(7), 961-967 (2012-12-18)
A systematic review and meta-analysis were conducted to examine the efficacy and safety of pegylated interferon (peg-IFN) alfa-2a and peg-IFN alfa-2b plus ribavirin (RBV) in children and adolescents with chronic hepatitis C virus (HCV). Medline, Embase, and Cochrane Central Register
Fred Poordad et al.
The New England journal of medicine, 370(21), 1973-1982 (2014-04-15)
Interferon-containing regimens for the treatment of hepatitis C virus (HCV) infection are associated with increased toxic effects in patients who also have cirrhosis. We evaluated the interferon-free combination of the protease inhibitor ABT-450 with ritonavir (ABT-450/r), the NS5A inhibitor ombitasvir
Michael Manns et al.
Lancet (London, England), 384(9954), 1597-1605 (2014-08-01)
An unmet need exists for interferon-free and ribavirin-free treatments for chronic hepatitis C virus (HCV) infection. In this study, we assessed all-oral therapy with daclatasvir (NS5A replication complex inhibitor) plus asunaprevir (NS3 protease inhibitor) in patients with genotype 1b infection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service