Skip to Content
Merck
All Photos(1)

Documents

T1100000

Thiamphenicol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-threo-2,2-Dichloro-N-(β-hydroxy-α-[hydroxymethyl]-4-[methylsulfonyl]phenethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15Cl2NO5S
CAS Number:
Molecular Weight:
356.22
Beilstein:
2819542
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thiamphenicol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CS(=O)(=O)c1ccc(cc1)[C@@H](O)[C@@H](CO)NC(=O)C(Cl)Cl

InChI

1S/C12H15Cl2NO5S/c1-21(19,20)8-4-2-7(3-5-8)10(17)9(6-16)15-12(18)11(13)14/h2-5,9-11,16-17H,6H2,1H3,(H,15,18)/t9-,10-/m1/s1

InChI key

OTVAEFIXJLOWRX-NXEZZACHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thiamphenicol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Malvy et al.
Intensive care medicine, 18(8), 485-487 (1992-01-01)
A 27-year-old woman, without compromised immunodefenses, experienced a Listeria meningoencephalitis, with brainstem symptoms. The identified agent exhibited poor susceptibility to usual effective antibiotics, except for penicillins. Knowledge of past history of an allergic reaction to beta-lactam antibiotics lead to appropriate
[Toxicity of chloramphenicol and thiamphenicol (CAP and TAP)].
G Keiser
Haematology and blood transfusion, 21, 179-195 (1978-01-01)
J Raymond et al.
Medecine tropicale : revue du Corps de sante colonial, 64(1), 33-38 (2004-07-01)
Streptococcus pneumoniae and Haemophilus influenzae are the two main pathogens responsible for bacterial respiratory tract infections. Their antimicrobial susceptibility to antibiotics like beta-lactams, macrolides or fluoroquinolones has been largely studied, while it remains less known to other antibiotics like thiamphenicol
Chloramphenicol and thiamphenicol.
J Fuglesang et al.
Antibiotics and chemotherapy, 31, 1-21 (1982-01-01)
[Chloramphenicol and thiamphenicol].
L Mariani
La Clinica terapeutica, 86(3), 195-218 (1978-08-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service