Skip to Content
Merck
All Photos(1)

Key Documents

PHL80378

Canadine

phyproof® Reference Substance

Synonym(s):

Tetrahydroberberine, 5,8,13,13a-Tetrahydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-alpha]quinolizine, Canadine, THB, Tetrahydroumbellatine, Xanthopuccine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4
CAS Number:
Molecular Weight:
339.39
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

SMILES string

COc1ccc2CC3N(CCc4cc5OCOc5cc34)Cc2c1OC

InChI

1S/C20H21NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,8-9,16H,5-7,10-11H2,1-2H3

InChI key

VZTUIEROBZXUFA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Dopamine receptor antagonist; nicotinic acetylcholine receptor (α4β2, α4β4, α7, α1β, 1γδ) modulator

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manead Khin et al.
The Yale journal of biology and medicine, 93(2), 265-275 (2020-07-02)
Hydrastis canadensis, commonly known as goldenseal, is a botanical native to the southeastern United States that has been used for the treatment of infection. The activity of goldenseal is often attributed to the presence of alkaloids (cyclic, nitrogen-containing compounds) present
Ha Gyeong Kim et al.
Pharmaceutics, 12(10) (2020-09-30)
In our previous study, Hwang-Ryun-Hae-Dok-Tang, which contains berberine (BBR) as a main active ingredient, inhibited cytochrome P450 (CYP) 2D6 in a quasi-irreversible manner. However, no information is available on the detailed mechanism of BBR-induced CYP2D6 inhibition. Thus, the present study
Kai Zhang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1165, 122523-122523 (2021-01-27)
Primary dysmenorrhea is a prevalent gynecological disorder that severely affects the quality of life in women. Yuanhu Zhitong oral liquid (YZOL) is a standardized herbal preparation frequently used in clinical practice and is a promising alternative therapy for primary dysmenorrhea.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service