Skip to Content
Merck
All Photos(1)

Key Documents

73490

Supelco

Nitron

for spectrophotometric det. of nitrate and perchlorate, ≥97.0%

Synonym(s):

1,4-Diphenyl-3-phenylamino-1,2,4-triazolium hydroxide inner salt, 3,5,6-Triphenyl-2,3,5,6-tetraazabicyclo[2.1.1]hex-1-ene, 4,5-Dihydro-2,4-diphenyl-5-(phenylimino)-1H-1,2,4-triazolium hydroxide inner salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16N4
CAS Number:
Molecular Weight:
312.37
Beilstein:
3913543
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥97.0% (NT)
≥97.0%

form

solid

quality

for spectrophotometric det. of nitrate and perchlorate

technique(s)

UV/Vis spectroscopy: suitable

impurities

≤3% water

mp

189-190 °C (dec.) (lit.)

SMILES string

[N-]1N(C=[N+](c2ccccc2)\C1=N\c3ccccc3)c4ccccc4

InChI

1S/C20H16N4/c1-4-10-17(11-5-1)21-20-22-24(19-14-8-3-9-15-19)16-23(20)18-12-6-2-7-13-18/h1-16H

InChI key

CWGBFIRHYJNILV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

For spectrophotometric determination of nitrate and perchlorate

Other Notes

Reagent for the precipitation of anions; reagent for the spectrophotometric determination of nitrate and perchlorate by a difference method; Reagent for the determination of traces of gold

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 24, 439-439 (1979)
Mikrochimica Acta, 758-758 (1972)
S. Shahine et al.
Fresenius Journal of Analytical Chemistry, 276, 380-380 (1975)
Mikrochimica Acta, 2, 75-75 (1976)
N. Ganchev et al.
Mikrochimica Acta, 476-476 (1971)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service