Skip to Content
Merck
All Photos(1)

Key Documents

62963

Sigma-Aldrich

L-α-Lysophosphatidylcholine

from soybean, ≥98.0% (10 mg phospholipid per ml CHCl3, TLC)

Synonym(s):

3-sn-Lysophosphatidylcholine, L-α-Lysolecithin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51321705
NACRES:
NA.25

biological source

soybean

Quality Level

description

zwitterionic

Assay

≥98.0% (10 mg phospholipid per ml CHCl3, TLC)

concentration

10 mg/mL in chloroform

density

1.47 g/mL at 20 °C (lit.)

lipid type

phosphoglycerides

shipped in

wet ice

storage temp.

−20°C

Other Notes

primarily C18 unsaturated fatty acids

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service