Skip to Content
Merck
All Photos(1)

Key Documents

61506

Supelco

L-Threonine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(2S,3R)-2-Amino-3-hydroxybutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
119.12
Beilstein:
1721646
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

256 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

C[C@@H](O)[C@H](N)C(O)=O

InChI

1S/C4H9NO3/c1-2(6)3(5)4(7)8/h2-3,6H,5H2,1H3,(H,7,8)/t2-,3+/m1/s1

InChI key

AYFVYJQAPQTCCC-GBXIJSLDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Ching Liu et al.
Molecular and cellular biology, 34(1), 57-70 (2013-10-30)
In budding yeast (Saccharomyces cerevisiae), the cell cycle-dependent telomere elongation by telomerase is controlled by the cyclin-dependent kinase 1 (Cdk1). The telomere length homeostasis is balanced between telomerase-unextendable and telomerase-extendable states that both require Cdc13. The recruitment of telomerase complex
Na Wang et al.
Cancer, 120(19), 2965-2979 (2014-06-06)
The telomerase reverse transcriptase (TERT) promoter mutations C228T and C250T have been found in many malignancies, including in thyroid carcinomas. However, it is unclear how early these mutations occur in thyroid tumorigenesis. The study included primary tumors from 58 patients
Anne-Sophie Van Rompuy et al.
Journal of neuropathology and experimental neurology, 73(2), 159-174 (2014-01-16)
Mutations in the parkin gene are the most common cause of early-onset autosomal recessive Parkinson disease (PD). The pathogenic mechanisms of how parkin mutations lead to the development of PD are not fully understood. Studies of cell cultures and of
Patricia Seoane-Collazo et al.
Endocrinology, 155(5), 1679-1689 (2014-02-13)
Nicotine, the main addictive component of tobacco, promotes body weight reduction in humans and rodents. Recent evidence has suggested that nicotine acts in the central nervous system to modulate energy balance. Specifically, nicotine modulates hypothalamic AMP-activated protein kinase to decrease
C Chaussain et al.
Journal of dental research, 93(4), 360-365 (2014-02-04)
In the literature, the enamelin gene ENAM has been repeatedly designated as a possible candidate for caries susceptibility. Here, we checked whether ENAM variants could increase caries susceptibility. To this aim, we sequenced coding exons and exon-intron boundaries of ENAM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service