Skip to Content
Merck
All Photos(1)

Key Documents

SLTEST000

Millipore

Integrity Test Kit for small volume devices

For bubble-point testing of small filter units.

Synonym(s):

Integrity Test Kit, Small Volume Integrity Kit, Small Volume Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151819
eCl@ss:
32031690
NACRES:
NB.22

Quality Level

description

for use with Integritest Exacta Automatic Filter Integrity Test Instrument

sterility

non-sterile

pore size

5.0 μm pore size

compatibility

for use with Integritest® 4

shipped in

ambient

General description

•Convenient bubble-point testing

•For vented and non-vented devices

•Tests wide range of pore sizes and membrane types

Millipore membrane filter units can be integrity-tested using the bubble point test method. Small, syringe-driven membrane filter units with 15 cm2 filtration area, such as Millex and Sterivex units, can be conveniently tested using the syringe-style integrity tester.

Legal Information

INTEGRITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Integrity Test Kit verifies small volume filter units' integrity, enhancing filtration reliability in lab processes.

Integrity Test Kit verifies small volume filter units' integrity, enhancing filtration reliability in lab processes.

Integrity Test Kit verifies small volume filter units' integrity, enhancing filtration reliability in lab processes.

Integrity Test Kit verifies small volume filter units' integrity, enhancing filtration reliability in lab processes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service