Skip to Content
Merck
All Photos(4)

Key Documents

MABF222

Sigma-Aldrich

Anti-IKKa (p45) Antibody, clone 881H3

clone 881H3, from mouse

Synonym(s):

Inhibitor of nuclear factor kappa-B kinase subunit alpha, I-kappa-B kinase alpha, IKK-A, IKK-alpha, IkBKA, IkappaB kinase, Conserved helix-loop-helix ubiquitous kinase, I-kappa-B kinase 1, IKK1, Nuclear factor NF-kappa-B inhibitor kinase alpha, NFKBIKA,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

881H3, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CHUK(1147)

General description

The protein named Inhibitor of nuclear factor kappa-B kinase subunit alpha, or alternatively I-kappa-B kinase alpha, IKK-A, IKK-alpha, IKKa (p45), IkBKA, IkappaB kinase, I-kappa-B kinase 1, IKK1, NF-kappa-B inhibitor kinase alpha, NFKBIKA, or transcription factor 16 (TCF-16) and encoded by the human gene CHUK/IKKA/TCF16 is a serine kinase that plays an essential role in the NF-kappa-B signaling pathway. IKKa(p45) acts as part of a complex that phosphorylates inhibitors of the NF-kappa-B. This allows then the polyubiquitination of the inhibitors and their subsequent degradation. Free NF-kappa-B is translocated into the nucleus and performs its gene activation functions governing everything from the immune response to growth and cell survival. IKKa(p45) can be found expressed in most tissues and localizes to both the cytoplasm and nucleus depending upon the microenvironment. Defects in IKKa(p45) are associated with a lethal fetal syndrome called by Cocoon syndrome where the fetus has multiple malformations including limbs bound to the trunk and encased under the skin.

Specificity

This antibody preferentially binds both the p45 fragment of IKKa (~45 kDa) as well as the full-length non-denatured form (~80-85 kDa). This antibody shows better preferential binding to the p45 fragment by immunoprecipitation, and detects both forms of IKKa by western blot.

Immunogen

Conjugated linear peptide corresponding to human IKKa (p45).

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected IKKa (p45) in human kidney and tonsil tissues.

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated IKKa (p45) from HCT116 cell lysate (Margalef, P., et al. (2012). Cell Rep. 2(40):840-854).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected IKKa (p45) in human intestinal mucosa, adenoma, and carcinoma tissues (Margalef, P., et al. (2012). Cell Rep. 2(40):840-854.).

Western Blotting Analysis: A previous lot of this antibody detected the truncated p45-IKKa form observed at ~45 kDa in WT MEF cell lysate, but less so in the KO MEFs, and the full length non-denatured protein at ~80-85 kDa. A non-specific band at ~75 kDa is detected.

Immunoprecipitation Analysis: A previous lot of this antibody immunoprecipiated the truncated p45-IKKa form observed at ~45 kDa.
Research Category
Inflammation & Immunology
Research Sub Category
Inflammation & Autoimmune Mechanisms
This Anti-IKKa (p45) Antibody, clone 881H3 is validated for use in western blotting, IHC (Paraffin) & IP for the detection of IKKa (p45).

Quality

Evaluated by Western Blotting in HCT116 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected IKKa (p45) in 10 µg of HCT116 cell lysate.

Target description

~80-85 kDa & ~45 kDa observed. The full length non-denatured protein may be observed at ~80-85 kDa, whereas the truncated p45-IKKa fragment may be observed at ~45 kDa in some lysates. A non-specific band at ~75 kDa may also be observed in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlota Colomer et al.
Molecular cell, 75(4), 669-682 (2019-07-16)
Phosphorylated IKKα(p45) is a nuclear active form of the IKKα kinase that is induced by the MAP kinases BRAF and TAK1 and promotes tumor growth independent of canonical NF-κB signaling. Insights into the sources of IKKα(p45) activation and its downstream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service