Skip to Content
Merck
All Photos(1)

Key Documents

LSKMAGL10

Millipore

PureProteome Albumin Magnetic Beads

The PureProteome Albumin Magnetic Beads are conjugated to an antibody specific for human serum albumin.

Synonym(s):

Albumin Magnetic Beads, Protein Bead Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
eCl@ss:
32160405
NACRES:
NA.56

packaging

pkg of 10 mL

manufacturer/tradename

PureProteome

technique(s)

depletion: suitable (serum)
protein purification: suitable

particle size

10 μm

shipped in

wet ice

storage temp.

2-8°C

General description

The PureProteome Albumin Magnetic Beads are conjugated to an antibody specific for human serum albumin. These magnetic beads provide a rapid, scalable, and reproducible means to deplete >98% of albumin from serum and plasma samples, facilitating the detection and analysis of proteins of interest. For efficient depletion, the PureProteome Magnetic Beads must be used with the PureProteome Magnetic Stand.

Analysis Note

Depletion: >98% depletion of Albumin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service