Skip to Content
Merck
All Photos(1)

Key Documents

AP184B

Sigma-Aldrich

Donkey Anti-Sheep IgG Antibody, biotin-SP conjugate, Species Adsorbed

Chemicon®, from donkey

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

biotin conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

sheep

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Sheep IgG (H & L)

F/P RATIO:

9.1 ug./mg.; 3.4 miles FITC per mole IgG

Application

Detect Donkey Sheep IgG using this Donkey anti-Sheep IgG Antibody, biotin-SP conjugate, Species Adsorbed validated for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
Suggested dilution for most applications: 1:50-1:200.

Optimal working dilutions must be determined by the end user.

Physical form

Lyophilized. Buffer = 0.02 M Sodium Phosphate, 0.25 M NaCl, pH 7.6, with 15 mg/mL BSA and 0.1% sodium azide.

RECONSTITUTION:

Reconstitute with 0.4ml distilled water.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Iwasa et al.
The Journal of veterinary medical science, 61(6), 621-629 (1999-07-29)
The present study showed the acetylcholinesterase (AChE) activity, and neurofilament protein (NFP), catecholamine-synthesizing enzymes, dopamine beta-hydroxylase (DBH) and phenylethanolamine N-methyltransferase (PNMT) immunoreactivities in the mouse adrenal gland during postnatal development. From birth to postnatal-1-day, AChE activity was weakly and diffusely
Yaqing Li et al.
Nature medicine, 23(6), 733-741 (2017-05-02)
Blood vessels in the central nervous system (CNS) are controlled by neuronal activity. For example, widespread vessel constriction (vessel tone) is induced by brainstem neurons that release the monoamines serotonin and noradrenaline, and local vessel dilation is induced by glutamatergic
Isabel Pérez-Santos et al.
Cerebral cortex (New York, N.Y. : 1991), 31(9), 4115-4139 (2021-05-19)
Noradrenaline (NA) in the thalamus has important roles in physiological, pharmacological, and pathological neuromodulation. In this work, a complete characterization of NA axons and Alpha adrenoceptors distributions is provided. NA axons, revealed by immunohistochemistry against the synthesizing enzyme and the
Yaqing Li et al.
Journal of neurophysiology, 111(1), 145-163 (2013-09-27)
Spinal cord transection leads to elimination of brain stem-derived monoamine fibers that normally synthesize most of the monoamines in the spinal cord, including serotonin (5-hydroxytryptamine, 5-HT) synthesized from tryptophan by enzymes tryptophan hydroxylase (TPH, synthesizing 5-hydroxytryptophan, 5-HTP) and aromatic l-amino
Inbar Zohar et al.
The European journal of neuroscience, 43(4), 590-600 (2015-12-17)
It is still not clear whether the selective serotonin reuptake inhibitors frequently prescribed to depressed pregnant women improve the behavioural outcome in their children. The current study investigated whether administration of citalopram to pregnant rats could prevent anxiety and depressive-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service