Skip to Content
Merck
All Photos(1)

Key Documents

5.05026

Sigma-Aldrich

DNQX Disodium Salt

Synonym(s):

DNQX Disodium Salt, AMPA Glutamate Receptor Antagonist, DNQX, Kainate Glutamate Receptor Antagonist, DNQX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H2N4Na2O6
CAS Number:
Molecular Weight:
296.10
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark brown

solubility

H2O: 100 mM

storage temp.

2-8°C

InChI

1S/C8H4N4O6/c13-7-8(14)10-4-2-6(12(17)18)5(11(15)16)1-3(4)9-7/h1-2H,(H,9,13)(H,10,14)

InChI key

RWVIMCIPOAXUDG-UHFFFAOYSA-N

General description

A potent competitive antagonist selective for AMPA/kainate glutamate receptors (IC50 = 0.19-1.66 µM and 0.35 µM for AMPA and GluK2 receptors, respectively). Routinely used as a tool to distinguish the role of neurotransmissions mediated by AMPA/kainate and NMDA receptors in studying brain functions and disorders.

Biochem/physiol Actions

Primary Target
AMPA
Secondary Target
GluK2
Target IC50: 0.19 to 1.66 uM

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, S., et al. 2010. J. Neurophysiol.103, 1728.
Honor, T., et al. 1998. Science.241, 701.
Watkins, J., et al. 1990. Adv. Exp. Med. Biol.268, 49.
Davies, S., et al. 1990. Int. Rev. Neurobiol.32, 28.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service