Skip to Content
Merck
All Photos(1)

Key Documents

444251-M

Millipore

MMP-2/MMP-9 Inhibitor III

The MMP-2/MMP-9 Inhibitor III controls the biological activity of MMP-2/MMP-9. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-2/MMP-9 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H71N13O14S2
Molecular Weight:
1166.33
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 5 mg/mL

storage temp.

−20°C

General description

A cyclic peptide that acts as a potent inhibitor of MMP-2 (gelatinase A; IC50 = 10 µM) and MMP-9 (gelatinase B; IC50 = 10 µM). Inhibits the migration of human endothelial cells and tumor cells, and prevents tumor growth and invasion in animal models.
A hydrophobic cyclic peptide that acts as a potent inhibitor of MMP-2 and MMP-9 (IC50 = 10 µM). Inhibits the migration of human endothelial cells and tumor cells, and prevents tumor growth and invasion in animal models. Also, phage that display this peptide specifically targets angiogenic blood vessels in vivo.

Biochem/physiol Actions

Target IC50: 10 µM against MMP-2 and MMP-9

Warning

Toxicity: Standard Handling (A)

Sequence

H-Cys¹-Thr-Thr-His-Trp-Gly-Phe-Thr-Leu-Cys¹⁰-OH (cyclic: 1→ 10)

Other Notes

Medina, O.P., et al. 2001. Cancer Res.61, 3978.
Koivunen, E., et al. 1999. Nat. Biotechnology 17, 768.

Legal Information

Sold under license of U.S. Patent 6,624,144.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service