Skip to Content
Merck
All Photos(1)

Key Documents

05-886R

Sigma-Aldrich

Anti-phospho-STAT5A/B (Tyr694/699) Antibody, clone 94-10C-9-10C-2, rabbit monoclonal

clone 94-10C-9-10C-2, from rabbit

Synonym(s):

signal transducer and activator of transcription 5A/B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

94-10C-9-10C-2, monoclonal

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr694/pTyr699)

Gene Information

human ... STAT5A(6776)

General description

STAT (Signal Transducers and Activators of Transcription) are a family of conserved transcription factors that transduce high-fidelity signals for the cytokine family of ligands and receptors as the principle substrates of JAK kinases (1,2). In resting cells, the STATs exist in an inactive state in the cytoplasm until activated. Activators include cytokines, growth factors, and some peptides. These activate JAK kinases, which when activated, bind to the STAT SH2 (Src homology domain-2) domain resulting in phosphorylation of STAT (1). Upon activation, STAT1, -3, -4, -5A, and -5B all form homodimers. STAT1 and STAT2, as well as STAT1 and STAT3 form heterodimers, and several STATs form tetramers, depending on the variables of the activating ligand (1). After dimerization, the STATs accumulate in the nucleus by way of nuclear transport (1). STAT5A and STAT5B appear to be most important to the function of growth hormone and prolactin and play critical roles in milk production (1,2,3). The phosphorylation of Tyrosine 694/699 is a hallmark of activated STAT5.

Specificity

Recognizes phosphorylated STAT5A/B atTyr694/Tyr699.

Immunogen

Epitope: Tyr694/Tyr699
KLH-conjugated linear peptide corresponding to phosphorylated human STAT5A/B at and around Tyr694/Tyr699.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Transcription Factors
Use Anti-phospho-STAT5A/B (Tyr694/699) Antibody, clone 94-10C-9-10C-2 (rabbit monoclonal antibody) validated in WB to detect phospho-STAT5A/B (Tyr694/699) also known as signal transducer & activator of transcription 5A/B.

Quality

Evaluated by Western Blot in U937 cell lysate.

Western Blot Analysis: 0.1 µg/ml of this antibody detected STAT5 A/B on 10 µg of U937 cell lysate.

Target description

95 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit monoclonal IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide and 30% Glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
GMCSF treated and untreated U937 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna M Schoepf et al.
European journal of medicinal chemistry, 216, 113285-113285 (2021-03-05)
The development of resistance poses a serious problem in the therapy of cancer due to the necessity of a multiple-drug and unlimited treatment of affected patients. In chronic myeloid leukemia (CML), the introduction of imatinib has revolutionized the therapy. The
Johannes W Bigenzahn et al.
Molecular & cellular proteomics : MCP, 15(3), 1139-1150 (2016-03-05)
Tandem affinity purification-mass spectrometry (TAP-MS) is a popular strategy for the identification of protein-protein interactions, characterization of protein complexes, and entire networks. Its employment in cellular settings best fitting the relevant physiology is limited by convenient expression vector systems. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service