Skip to Content
Merck
All Photos(1)

Key Documents

P-115

Supelco

PEth 16:0/18:2 solution

1.0 mg/mL in methanol (as free phosphate), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O8P · NC16H36
Molecular Weight:
942.42
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free phosphate)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−70°C

SMILES string

CCOP(OC[C@H](OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C39H73O8P.C16H36N/c1-4-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-39(41)47-37(36-46-48(42,43)45-6-3)35-44-38(40)33-31-29-27-25-23-21-18-16-14-12-10-8-5-2;1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4/h13,15,19-20,37H,4-12,14,16-18,21-36H2,1-3H3,(H,42,43);5-16H2,1-4H3/q;+1/p-1/b15-13-,20-19-;/t37-;/m1./s1

InChI key

BVQILUQKSJVANH-CZRMIBBWSA-M

General description

PEth 16:0/18:2, also known as phosphatidylethanol 16:0/18:2, is a sensitive and specific biomarker for alcohol use and abuse. Since the detection window of PEth 16:0/18:2, in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/18:2, levels is routinely performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing. PEth 16:0/18:2 has also been investigated as a biomarker to confirm prenatal alcohol exposure (PAE).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Jeffery et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 35(6), 663-668 (1987-06-01)
Thyroidectomy has a dramatic effect on rat muscle, greatly increasing the number of Type I fibers and the concentrations of carbonic anhydrase III (CAIII) in the muscle. Carbonic anhydrase III is not confined to the Type I fibers, as was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service