Skip to Content
Merck
All Photos(2)

Key Documents

850358C

Avanti

16:1 (Δ9-Cis) PC

1,2-dipalmitoleoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1,2-di-(9Z-hexadecenoyl)-sn-glycero-3-phosphocholine; PC(16:1(9Z)/16:1(9Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H76NO8P
CAS Number:
Molecular Weight:
730.01
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850358C-25mg)
pkg of 2 × 4 mL (850358C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850358C

concentration

10 mg/mL (850358C-25mg)
25 mg/mL (850358C-200mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

16:1 (Δ9-Cis) PC (1,2-dipalmitoleoyl-sn-glycero-3-phosphocholine) is a lipid derivative having a phosphocholine backbone. It also has two palmitic acid chains each having one cis double bond at 9th carbon. It is a double mono-unsaturated lipid.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

16:1 (Δ9-Cis) PC (1,2-dipalmitoleoyl-sn-glycero-3-phosphocholine) has been used in liposome preparation. It may be used to prepare giant unilamellar vesicles (GUVs) and multilamellar vesicles (MLVs).

Biochem/physiol Actions

Phosphatidylcholine (PC) forms a hydrophobic surface in the mucus by acting as a surfactant resulting in the inhibition of penetrance of bacteria.

Packaging

5 mL Clear Glass Sealed Ampule (850358C-200mg)
5 mL Clear Glass Sealed Ampule (850358C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bilayer thickness and curvature influence binding and insertion of a pHLIP peptide
Karabadzhak AG, et al
Biophysical Journal (2018)
Parian AM
Integrative Medicine (2018)
Solubilization of lipids and lipid phases by the styrene-maleic acid copolymer
<BIG><BIG>Pardo JJD, et al</BIG></BIG>
European Biophysics Journal (2017)
Łukasz Syga et al.
Chembiochem : a European journal of chemical biology, 21(9), 1320-1328 (2019-12-10)
Attachment of lipophilic groups is an important post-translational modification of proteins, which involves the coupling of one or more anchors such as fatty acids, isoprenoids, phospholipids, or glycosylphosphatidyl inositols. To study its impact on the membrane partitioning of hydrophobic peptides
Miranda L Schmidt et al.
Biochimica et biophysica acta. Biomembranes, 1862(5), 183196-183196 (2020-01-21)
Lipid bilayers form the basis of cell membranes and the phase behaviour of the membrane has been linked to proper cell function. Model membranes composed of relatively simple mixtures of phospholipids and cholesterol can already exhibit complex phase behaviour. Specifically

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service