Skip to Content
Merck
All Photos(3)

Key Documents

840024P

Avanti

Soy PE

Avanti Research - A Croda Brand 840024P, powder

Synonym(s):

L-α-phosphatidylethanolamine (Soy)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (840024P-200mg)
pkg of 1 × 25 mg (840024P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840024P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

InChI

1S/C39H74NO8P/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-39(42)48-37(36-47-49(43,44)46-34-33-40)35-45-38(41)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h11,13,17-18,37H,3-10,12,14-16,19-36,40H2,1-2H3,(H,43,44)/b13-11-,18-17-/t37-/m1/s1

InChI key

HBZNVZIRJWODIB-NHCUFCNUSA-N

General description

L-α-phosphatidylethanolamine is a naturally occurring aminophospholipid in soy lecithin. Soy PE is one of the important components of biological membranes.

Application

Soy PE has been used in lipid kinase assays.

Biochem/physiol Actions

Soy PE plays a vital role in structural integrity and functionality of cells. It possesses anti-oxidative properties.

Packaging

5 mL Clear Glass Sealed Ampule (840024P-200mg)
5 mL Clear Glass Sealed Ampule (840024P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and quantification of phosphatidylethanolamine-derived glucosylamines and aminoketoses from human erythrocytes?influence of glycation products on lipid peroxidation
Carmen MBU, et al.
Archives of Biochemistry and Biophysics, 391(2), 245-254 (2001)
Quercetin increases antioxidant activity of soy lecithin in a triolein model system
Ramadan MF
LWT--Food Science and Technology, 41(4) (2008)
Design and structural characterization of potent and selective inhibitors of phosphatidylinositol 4 kinase IIIbeta
Rutaganira FU, et al.
Journal of Medicinal Chemistry, 59(5), 1830-1839 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service