Skip to Content
Merck
All Photos(2)

Key Documents

M25406

Sigma-Aldrich

2-Methoxypyridine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
Beilstein:
108189
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.503 (lit.)

bp

142 °C (lit.)

density

1.038 g/mL at 25 °C (lit.)

SMILES string

COc1ccccn1

InChI

1S/C6H7NO/c1-8-6-4-2-3-5-7-6/h2-5H,1H3

InChI key

IWTFOFMTUOBLHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenshuai Dai et al.
ChemistryOpen, 8(7), 840-851 (2019-07-16)
Multiple approaches reveal the strong effects of a positive charge introduced by protonation or ionization on the conformation of o-heteroaromatic ethers and amides. The ethers and amides containing an ortho-N heteroatom are syn-preferring while those containing an ortho-O or ortho-S
Saira Khalid et al.
Environmental science and pollution research international, 23(20), 20646-20660 (2016-07-30)
Application of chlorpyrifos (CP) has increased its environmental concentration. Increasing CP concentration has increased chances of adverse health effects. Its removal from environment has attained researcher's attention. CP degrading bacterial strains were isolated from wastewater and agricultural soil. Finally, selected
Masazumi Tamura et al.
Nature communications, 6, 8580-8580 (2015-10-06)
Multidentate materials formed by simply mixing heterogeneous and homogeneous components are promising for construction of versatile active sites on the surface of heterogeneous compounds, however, to the best of our knowledge, there are no reports on such materials. Self-assembly of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service