Skip to Content
Merck
All Photos(1)

Key Documents

772445

Sigma-Aldrich

Fmoc-Ser[GalNAc(Ac)3-α-D]-OH

95%, for peptide synthesis

Synonym(s):

N-α-Fmoc-O-β-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-galactopyranosyl)-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H36N2O13
CAS Number:
Molecular Weight:
656.63
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

Fmoc-Ser[GalNAc(Ac)3-α-D]-OH, 95%

Assay

95%

form

solid

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

CC(O[C@@H]([C@H](COC(C)=O)O1)[C@H](OC(C)=O)[C@@H](NC(C)=O)[C@H]1OC[C@@H](C(O)=O)NC(OCC2C3=C(C=CC=C3)C4=C2C=CC=C4)=O)=O

InChI

1S/C32H36N2O13/c1-16(35)33-27-29(46-19(4)38)28(45-18(3)37)26(15-42-17(2)36)47-31(27)43-14-25(30(39)40)34-32(41)44-13-24-22-11-7-5-9-20(22)21-10-6-8-12-23(21)24/h5-12,24-29,31H,13-15H2,1-4H3,(H,33,35)(H,34,41)(H,39,40)/t25-,26-,27+,28-,29+,31-/m0/s1

InChI key

ORICVOOXZDVFIP-MIOLQQMZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service